1-Phenyl-1-propyne 95%

Supplier: Ambeed
Warning

Synonyms: 1-Propynylbenzene, Prop-1-yn-1-ylbenzene

A375679-1G A375679-5G
103826-566EA 23 USD
103826-566 103837-590 76840-880 76840-882
1-Phenyl-1-propyne 95%
1-Phenyl-1-propyne

Precaución: for Research Use Only

Formula: C₆H₅CCCH₃
MW: 116.16 g/mol
Boiling Pt: 182 °C (760 mm Hg)
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009272
CAS Number: 673-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR