Praziquantel 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Cyclohexylcarbonyl)-1,2,3,6,7,11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one, 2-(Cyclohexanecarbonyl)-2,3,6,7-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4(11bH)-one

A186621-100G A186621-10G A186621-10MG A186621-50MG A186621-25G A186621-5G A186621-1MG A186621-500G
103846-532EA 10 USD
103846-532 76721-030 76721-032 76721-034 76721-036 77302-038 77302-040 77600-512 77667-554
Praziquantel 98%
Praziquantel

Precaución: for Research Use Only

Formula: C₁₉H₂₄N₂O₂
MW: 312.41 g/mol
Boiling Pt: 544.1 °C (760 mm Hg)
Melting Pt: 136…138 °C
Flash Pt: 255 °C
Storage Temperature: Freezer
MDL Number: MFCD00058531
CAS Number: 55268-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR