Pravastatin sodium 98%

Supplier: Ambeed
Warning

Synonyms: (3R,5R)-3,5-Dihydroxy-7-((1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-(((S)-2-methylbutanoyl)oxy)-1,2,6,7,8,8a-hexahydronaphthalen-1-yl)heptanoic acid, sodium salt

A128695-100MG A128695-1MG A128695-250MG A128695-10MG A128695-50MG A128695-1G
103792-114EA 26 USD
103792-114 103799-330 76782-184 76782-186 76782-188 77298-314 77298-316 77566-502
Pravastatin sodium 98%
Pravastatin sodium

Precaución: For research use only.

Formula: C₂₃H₃₅NaO₇
Boiling Pt: 634.5 °C (760 mm Hg)
Melting Pt: 171.2…173 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00887601
CAS Number: 81131-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR