Potassium phenyltrifluoroborate 97%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro(phenyl)borate, Phenyltrifluoroboric acid potassium salt

A145448-100G A145448-10G A145448-250MG A145448-1G A145448-25G A145448-5G
103803-536EA 6 USD
103803-536 103841-622 77095-598 77095-600 77095-602 77316-918 77316-920 77316-922
Potassium phenyltrifluoroborate 97%
Potassium phenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₆H₅BF₃K
MW: 184.01 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD01318172
CAS Number: 153766-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR