Potassium trifluoro(oxetan-3-yl)borate 95%

Supplier: Ambeed
Warning

A125263-100MG A125263-250MG
103800-656EA 512 USD
103800-656 77571-508 77571-510
Potassium trifluoro(oxetan-3-yl)borate 95%
Potassium trifluoro(oxetan-3-yl)borate

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₃H₅BF₃O·K
Storage Temperature: Refrigerator
MDL Number: MFCD24843091
CAS Number: 1430219-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR