Potassium (2,2-dimethylprop-1-yl)trifluoroborate 95%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro(neopentyl)borate

A172655-250MG A172655-1G A172655-5G
103806-736EA 46 USD
103806-736 103845-028 76824-556 77082-232 77082-234
Potassium (2,2-dimethylprop-1-yl)trifluoroborate 95%
Potassium (2,2-dimethylprop-1-yl)trifluoroborate

Precaución: for Research Use Only

Formula: C₅H₁₁BF₃·K
MW: 178.045 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09271780
CAS Number: 1150655-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR