Potassium isopropyltrifluoroborate 95%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro(isopropyl)borate

A279874-10G A279874-250MG A279874-1G A279874-25G A279874-5G
103813-490EA 6 USD
103813-490 103853-434 76827-538 76827-540 77093-360 77093-362 77093-364
Potassium isopropyltrifluoroborate 95%
Potassium isopropyltrifluoroborate

Precaución: for Research Use Only

Formula: C₃H₇BF₃K
Melting Pt: >300 °C
Storage Temperature: Freezer
MDL Number: MFCD04112718
CAS Number: 1041642-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR