Potassium trifluoro(6-fluoropyridin-3-yl)borate 95%

Supplier: Ambeed
Warning

A172329-10G A172329-250MG A172329-1G A172329-25G A172329-5G
103806-718EA 12 USD
103806-718 103845-002 76985-496 76985-498 77301-196 77301-198 77301-200
Potassium trifluoro(6-fluoropyridin-3-yl)borate 95%
Potassium trifluoro(6-fluoropyridin-3-yl)borate

Precaución: for Research Use Only

Formula: C₅H₃BF₄KN
Storage Temperature: Ambient
MDL Number: MFCD09993004
CAS Number: 1111732-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR