Potassium(4-methoxyphenyl)trifluoroborate 97%

Supplier: Ambeed
Warning

Synonyms: (4-Methoxyphenyl)trifluoroboric acid potassium salt, Potassium (4-methoxyphenyl)trifluoroborate, Potassium trifluoro(4-methoxyphenyl)borate

A130679-250MG A130679-1G A130679-5G
103801-458EA 19 USD
103801-458 76718-934 77298-424 77313-912
Potassium(4-methoxyphenyl)trifluoroborate 97%
Potassium(4-methoxyphenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₇H₇BF₃KO
MW: 214.04
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD04112717
CAS Number: 192863-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR