Potassium(4-fluorophenyl)trifluoroborate 97%

Supplier: Ambeed
Warning

Synonyms: (4-Fluorophenyl)trifluoroboric acid potassium salt, Potassium trifluoro(4-fluorophenyl)borate

A128679-100G A128679-250MG A128679-1G A128679-5G
103801-172EA 9 USD
103801-172 77094-870 77094-872 77313-514 77313-516
Potassium(4-fluorophenyl)trifluoroborate 97%
Potassium(4-fluorophenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₆H₄BF₄K
MW: 202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318170
CAS Number: 192863-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR