Potassium(3-methoxyphenyl)trifluoroborate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Potassium trifluoro(3-methoxyphenyl)borate, (3-Methoxyphenyl)trifluoroboric acid potassium salt
A431065-250MG
A431065-1G
A431065-5G
103827-518EA
9
USD
103827-518
103838-584
76725-396
76842-276
76842-278
Potassium(3-methoxyphenyl)trifluoroborate 97%
Potassium(3-methoxyphenyl)trifluoroborate
Precaución: for Research Use Only
Formula:
C₇H₇BF₃KO MW: 214.04 g/mol Melting Pt: 178…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD04039954 CAS Number: 438553-44-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...