Potassium(3-fluorophenyl)trifluoroborate 97%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro(3-fluorophenyl)borate, (3-Fluorophenyl)trifluoroboric acid potassium salt

A210588-10G A210588-250MG A210588-1G A210588-100G
103818-022EA 15 USD
103818-022 103848-716 76721-672 76721-674 77087-096 77879-306
Potassium(3-fluorophenyl)trifluoroborate 97%
Potassium(3-fluorophenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₆H₄BF₄K
MW: 202
Melting Pt: 188…193 °C
Storage Temperature: Ambient
MDL Number: MFCD06659938
CAS Number: 267006-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR