Potassium tetrachloroplatinate(II) 98%

Supplier: Ambeed
Danger

A128488-10G A128488-250MG A128488-1G A128488-25G A128488-5G
103801-142EA 34 USD
103801-142 76718-868 76718-870 77298-306 77298-308 77313-462
Potassium tetrachloroplatinate(II) 98%
Potassium tetrachloroplatinate(II)

Precaución: for Research Use Only

Formula: Cl₄K₂Pt
Melting Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00011378
CAS Number: 10025-99-7
UN: 3288
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR