potassium trimethylacetate 95%

Supplier: Ambeed
Warning

Synonyms: 2,2-Dimethylpropionic acid potassium salt, Pivalic acid potassium salt, Potassium trimethylacetate, Potassium Pivalate, Trimethylacetic acid potassium salt, Potassium 2,2-Dimethylpropionate

A301804-100G A301804-10G A301804-500G A301804-25G A301804-5G
103810-018EA 351 USD
103810-018 103835-464 76827-872 76827-874 76827-876 76838-162 77305-828
potassium trimethylacetate 95%
potassium trimethylacetate

Precaución: for Research Use Only

Formula: C₅H₉KO₂
MW: 140.223 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00671345
CAS Number: 19455-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR