Potassium 2-phenylethyltrifluoroborate 97%

Supplier: Ambeed
Warning

Synonyms: Potassium Trifluoro(phenethyl)borate, Potassium Trifluoro(2-phenylethyl)borate, Potassium Phenethyltrifluoroborate

A801445-250MG A801445-1G
103841-066EA 110 USD
103841-066 76728-570 76845-308
Potassium 2-phenylethyltrifluoroborate 97%
Potassium 2-phenylethyltrifluoroborate

Precaución: for Research Use Only

Formula: C₈H₉BF₃K
Melting Pt: 173…175 °C
Storage Temperature: Freezer
MDL Number: MFCD09039257
CAS Number: 329976-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR