Potassium ethyltrifluoroborate 95%

Supplier: Ambeed
Warning

Synonyms: Potassium ethyltrifluoroboranuide

A132145-10G A132145-250MG A132145-1G
103801-692EA 15 USD
103801-692 77149-654 77149-656 77314-250
Potassium ethyltrifluoroborate 95%
Potassium ethyltrifluoroborate

Precaución: for Research Use Only

Formula: C₂H₅BF₃K
MW: 135.9656 g/mol
Melting Pt: 80…84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04112713
CAS Number: 44248-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR