Potassium-3,4-(methylenedioxy)phenyltrifluoroborate 97%

Supplier: Ambeed
Warning

Synonyms: Potassium benzo[d][1,3]dioxol-5-yltrifluoroborate

A128873-1G A128873-5G
103801-192EA 15 USD
103801-192 77313-548 77313-550
Potassium-3,4-(methylenedioxy)phenyltrifluoroborate 97%
Potassium 3,4-(methylenedioxy)phenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₇H₅BF₃KO₂
MW: 228.0200 g/mol
Melting Pt: 236…242 °C
Storage Temperature: Ambient
MDL Number: MFCD08276799
CAS Number: 871231-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR