Potassium (4-pyridyl)trifluoroborate 95%

Supplier: Ambeed
Warning

Synonyms: Potassium 4-Pyridyltrifluoroborate

A206834-1G A206834-25G A206834-5G A206834-250MG
103817-736EA 27 USD
103817-736 103848-416 77086-674 77086-676 77086-678 77152-336
Potassium (4-pyridyl)trifluoroborate 95%
Potassium (4-pyridyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₅H₄BF₃KN
MW: 184.9965 g/mol
Melting Pt: 45…50 °C
Storage Temperature: Ambient
MDL Number: MFCD09038543
CAS Number: 1111732-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR