Potassium-2,3,3-trimethyl-3H-indole-5-sulfonate 97%

Supplier: Ambeed
Warning

Synonyms: Potassium 2,3,3-trimethyl-3H-indole-5-sulphonate

A126736-100MG A126736-250MG A126736-1G
103792-100EA 150 USD
103792-100 103799-324 103800-856 76810-954 76810-956 77298-146
Potassium-2,3,3-trimethyl-3H-indole-5-sulfonate 97%
Potassium-2,3,3-trimethyl-3H-indole-5-sulfonate

Precaución: for Research Use Only

Formula: C₁₁H₁₂KNO₃S
MW: 277.39 g/mol
Melting Pt: 262…264 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07369684
CAS Number: 184351-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR