Potassium trifluoroacetate 98%

Supplier: Ambeed
Warning

Synonyms: Trifluoroacetic acid potassium salt, Potassium 2,2,2-trifluoroacetate

A146451-100G A146451-10G A146451-1G A146451-500G A146451-25G A146451-5G A146451-1000G
103808-676EA 93 USD
103808-676 103841-758 76719-534 76719-536 76719-538 76719-540 77299-520 77299-522 77317-082
Potassium trifluoroacetate 98%
Potassium trifluoroacetate

Precaución: for Research Use Only

Formula: C₂F₃KO₂
MW: 152.11 g/mol
Melting Pt: 143…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00013215
CAS Number: 2923-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR