Phthalimide potassium salt 98%
Supplier: Ambeed
|
Warning
|
A207437-100G
A207437-500G
A207437-1000G
A207437-25G
103809-432EA
27
USD
103809-432
76721-606
76721-608
77303-034
77303-036
Phthalimide potassium salt 98%
Phthalimide potassium salt
Precaución: for Research Use Only
Formula:
C₈H₄KNO₂ Melting Pt: >300 °C Flash Pt: 170.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005887 CAS Number: 1074-82-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...