Potassium(bromomethyl)trifluoroborate 95%

Supplier: Ambeed
Warning

Synonyms: (Bromomethyl)trifluoroboric acid potassium salt, Potassium (bromomethyl)trifluoroborate

A214216-100G A214216-10G A214216-500G A214216-1G A214216-25G A214216-5G
103818-378EA 5 USD
103818-378 103849-086 76721-810 76721-812 76721-814 77303-330 77303-332 77303-334
Potassium(bromomethyl)trifluoroborate 95%
Potassium(bromomethyl)trifluoroborate

Precaución: for Research Use Only

Formula: CH₂BBrF₃K
MW: 200.83 g/mol
Melting Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD09265154
CAS Number: 888711-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR