Potassium (4-cyanophenyl)trifluoroborate 97%

Supplier: Ambeed
Warning

A330304-10G A330304-250MG A330304-25G A330304-1G
103825-546EA 10 USD
103825-546 103836-506 76828-326 76828-328 76828-330 76839-484
Potassium (4-cyanophenyl)trifluoroborate 97%
Potassium (4-cyanophenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₇H₄BF₃KN
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD04115736
CAS Number: 850623-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR