Potassium(4-chlorophenyl)trifluoroborate 97%

Supplier: Ambeed
Warning

Synonyms: (4-Chlorophenyl)trifluoroboric acid potassium salt, Potassium (4-chlorophenyl)trifluoroborate

A303831-250MG A303831-1G A303831-5G A303831-10G
103814-396EA 7 USD
103814-396 103835-562 76723-660 76838-250 77305-888 77879-599
Potassium(4-chlorophenyl)trifluoroborate 97%
Potassium(4-chlorophenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₆H₄BClF₃K
MW: 218.45
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115770
CAS Number: 661465-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR