Potassium(4-bromophenyl)trifluoroborate 97%

Supplier: Ambeed
Warning

Synonyms: (4-Bromophenyl)trifluoroboric acid potassium salt, Potassium (4-bromophenyl)trifluoroborate, Potassium 4-bromophenyltrifluoroborate

A154497-100G A154497-10G A154497-1G A154497-25G
103804-648EA 6 USD
103804-648 103842-838 76719-876 76719-878 77318-566 77318-568
Potassium(4-bromophenyl)trifluoroborate 97%
Potassium(4-bromophenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₆H₄BBrF₃K
MW: 262.91 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD02093978
CAS Number: 374564-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR