Potassium(3-chlorophenyl)trifluoroborate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Potassium (3-chlorophenyl)trifluoroborate, (3-Chlorophenyl)trifluoroboric acid potassium salt
A320775-10G
A320775-250MG
A320775-1G
A320775-5G
103825-248EA
34
USD
103825-248
103836-180
76723-972
76723-974
76839-062
77306-224
Potassium(3-chlorophenyl)trifluoroborate 97%
Potassium(3-chlorophenyl)trifluoroborate
Precaución: for Research Use Only
Formula:
C₆H₄BClF₃K MW: 218.45 Melting Pt: 76…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115754 CAS Number: 411206-75-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...