Potassium(3-chloro-4-fluorophenyl)trifluoroborate 95%

Supplier: Ambeed
Warning

Synonyms: Potassium (3-chloro-4-fluorophenyl)trifluoroborate, (3-Chloro-4-fluorophenyl)trifluoroboric acid potassium salt

A219372-250MG A219372-1G A219372-5G
103818-784EA 24 USD
103818-784 103849-660 76721-928 76721-930 77088-214 77088-216
Potassium(3-chloro-4-fluorophenyl)trifluoroborate 95%
Potassium(3-chloro-4-fluorophenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₆H₃BClF₄K
MW: 236.44
Melting Pt: 234…244 °C
Storage Temperature: Ambient
MDL Number: MFCD04115752
CAS Number: 850623-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR