Potassium-3,5-bis(trifluoromethyl)phenyltrifluoroborate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Potassium [3,5-bis(trifluoromethyl)phenyl]trifluoroborate, Potassium (3,5-bis(trifluoromethyl)phenyl)trifluoroborate
A101607-10G
A101607-250MG
A101607-1G
A101607-5G
103796-044EA
13
USD
103796-044
103821-734
76717-556
76717-558
77297-162
77297-164
Potassium-3,5-bis(trifluoromethyl)phenyltrifluoroborate 97%
Potassium 3,5-bis(trifluoromethyl)phenyltrifluoroborate
Precaución: for Research Use Only
Formula:
C₈H₃BF₉K MW: 320.01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03701625 CAS Number: 166328-09-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...