Potassium-2,4-difluorophenyltrifluoroborate 95%

Supplier: Ambeed
Warning

Synonyms: Potassium (2,4-difluorophenyl)trifluoroborate

A127428-250MG A127428-1G A127428-5G
103800-962EA 40 USD
103800-962 76718-800 77298-208 77298-210
Potassium-2,4-difluorophenyltrifluoroborate 95%
Potassium 2,4-difluorophenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₆H₃BF₅K
MW: 219.99 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD07784375
CAS Number: 871231-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR