Pomalidomide 98%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-N-(2,6-dioxo-3-piperidyl)phthalimide, 4-Amino-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione

A203253-100G A203253-10G A203253-250MG A203253-1G A203253-5G
103817-454EA 8 USD
103817-454 103848-108 76825-614 76825-616 76825-618 77086-238 77086-240
Pomalidomide 98%
Pomalidomide

Precaución: for Research Use Only

Formula: C₁₃H₁₁N₃O₄
Boiling Pt: 582.9 °C (760 mm Hg)
Melting Pt: 315.5…317.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12756407
CAS Number: 19171-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR