Pivalamidine hydrochloride 97%

Supplier: Ambeed
Warning

A254703-100G A254703-10G A254703-250MG A254703-500G A254703-1G A254703-25G A254703-5G
103812-154EA 8 USD
103812-154 103852-008 76786-872 76786-874 76786-876 76786-878 77091-448 77091-450 77091-452
Pivalamidine hydrochloride 97%
Pivalamidine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂N₂
Melting Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00051988
CAS Number: 18202-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR