Piribedil 98%

Supplier: Ambeed
Warning

Synonyms: 2-[4-(1,3-Benzodioxol-5-ylmethyl)-1-piperazinyl]pyrimidine, 2-(4-(Benzo[d][1,3]dioxol-5-ylmethyl)piperazin-1-yl)pyrimidine

A254002-5G A254002-100MG A254002-10G A254002-250MG A254002-50MG
103812-120EA 13 USD
103812-120 103851-974 77091-412 77249-998 77250-000 77250-002 77250-004
Piribedil 98%
Piribedil

Precaución: for Research Use Only

Formula: C₁₆H₁₈N₄O₂
MW: 298.35 g/mol
Boiling Pt: 469.4 °C (760 mm Hg)
Flash Pt: 237.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00868264
CAS Number: 3605-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR