2-Piperidinecarboxamide 97%

Supplier: Ambeed
Warning

Synonyms: Piperidine-2-carboxamide

A192480-25G A192480-5G A192480-100G A192480-250MG
103816-444EA 16 USD
103816-444 103847-052 77084-924 77084-926 77111-820 77111-822
2-Piperidinecarboxamide 97%
2-Piperidinecarboxamide

Precaución: for Research Use Only

Formula: C₆H₁₂N₂O
MW: 128.18 g/mol
Melting Pt: 138…140 °C
Flash Pt: 142.3 °C
Storage Temperature: Ambient
MDL Number: MFCD05663784
CAS Number: 19889-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR