Glutarimide 98%

Supplier: Ambeed
Warning

Synonyms: Piperidine-2,6-dione

A100684-100G A100684-10G A100684-25G A100684-500G A100684-1G A100684-5G
103795-576EA 5 USD
103795-576 103821-270 76796-236 76796-238 76796-240 76796-242 77296-808 77296-810
Glutarimide 98%
Glutarimide

Precaución: for Research Use Only

Formula: C₅H₇NO₂
MW: 113.12 g/mol
Boiling Pt: 288 °C (760 mm Hg)
Melting Pt: 155…157 °C
Flash Pt: 152.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00006670
CAS Number: 1121-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR