Piperidin-3-ol 98%

Supplier: Ambeed
Danger

Synonyms: 3-Piperidinol, 3-Hydroxypiperidine

A682284-25G A682284-5G A682284-10G
103840-542EA 5 USD
103840-542 76844-744 76844-746 77293-152
Piperidin-3-ol 98%
Piperidin-3-ol

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 191.9 °C (760 mm Hg)
Melting Pt: 56…60 °C
Flash Pt: 100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014591
CAS Number: 6859-99-0
UN: 3263
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR