Pioglitazone ≥99% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: 5-(4-(2-(5-Ethylpyridin-2-yl)ethoxy)benzyl)thiazolidine-2,4-dione

A612927-1MG A612927-250MG A612927-50MG A612927-1G
103840-180EA 267 USD
103840-180 76775-318 76775-320 76775-322 76844-346
Pioglitazone ≥99% (by HPLC)
Pioglitazone

Precaución: for Research Use Only

Formula: C₁₉H₂₀N₂O₃S
MW: 356.44 g/mol
Boiling Pt: 575.4 °C (760 mm Hg)
Flash Pt: 301.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00865504
CAS Number: 111025-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR