4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane 97% stabilized

Supplier: Ambeed
Danger

Synonyms: Pinacol vinylboronate, Vinylboronic Acid Pinacol Ester

Stabilization: Stabilized with PTZ

A469455-10G A469455-25G A469455-5G
103828-008EA 10 USD
103828-008 103839-082 76789-552 76842-988 76842-990
4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane 97% stabilized
4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₈H₁₅BO₂
MW: 154.02 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00192492
CAS Number: 75927-49-0
UN: 3272
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR