Pimelic acid 98%

Supplier: Ambeed
Warning

Synonyms: Heptanedioic acid, 1,5-Pentanedicarboxylic acid, 1,7-Heptanedioic acid

A110325-100G A110325-10G A110325-25G A110325-500G A110325-5G A110325-1000G
103808-046EA 377 USD
103808-046 77107-898 77107-900 77107-902 77107-904 77107-906 77283-978
Pimelic acid 98%
Pimelic acid

Precaución: for Research Use Only

Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 212 °C (760 mm Hg)
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00004425
CAS Number: 111-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR