Pimecrolimus ≥98%
Supplier: Ambeed
|
Danger
|
Synonyms:
(3S,4R,5S,8R,9E,12S,14S,15R,16S,18R,19R,26aS)-3-[(1E)-2-[(1R,3R,4S)-4-Chloro-3-methoxycyclohexyl]-1-methylethenyl]-8-ethyl-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-14,16-dimethoxy-4,10,12,18-tetramethyl-15,19-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclotricosine-1,7,20,21(4H,23H)-tetrone
A944531-1G
A944531-5MG
A944531-1MG
103792-880EA
14
USD
103792-880
103799-742
77603-328
77603-330
77670-380
Pimecrolimus ≥98%
Pimecrolimus
Precaución: Keep in a dark place, sealed in dry, at 2…8°C
Boiling Pt:
866.1 °C (760 mm Hg) Flash Pt: 477.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00901792 CAS Number: 137071-32-0 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...