Pidotimod 98%

Supplier: Ambeed
Warning

Synonyms: (R)-3-[[(S)-5-Oxo-2-pyrrolidinyl]carbonyl]-1,3-thiazolidine-4-carboxylic Acid

A147731-100G A147731-100MG A147731-10G A147731-1G A147731-25G A147731-5G
103803-818EA 7 USD
103803-818 103841-926 76719-610 76719-612 76719-614 77317-324 77317-326 77317-328
Pidotimod 98%
Pidotimod

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₄S
MW: 244.27 g/mol
Melting Pt: 194…198 °C
Flash Pt: 354.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00867583
CAS Number: 121808-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR