Picolinamidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Picolinimidamide hydrochloride, 2-Amidinopyridine hydrochloride

A146124-100G A146124-10G A146124-500G A146124-1G A146124-25G A146124-5G
103803-632EA 5 USD
103803-632 103841-718 77095-626 77095-628 77095-630 77317-026 77317-028 77317-030
Picolinamidine hydrochloride 97%
Picolinamidine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₈ClN₃
MW: 157.60 g/mol
Melting Pt: 150…152 °C
Flash Pt: 99.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00052271
CAS Number: 51285-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR