Picolinic acid 97%

Supplier: Ambeed
Danger

Synonyms: Pyridine-2-carboxylic acid, 2-Picolinic acid, 2-Pyridinecarboxylic acid

A191920-100G A191920-10G A191920-500G A191920-1000G A191920-25G
103809-264EA 58 USD
103809-264 77029-844 77029-846 77029-848 77084-878 77302-272
Picolinic acid 97%
Picolinic acid

Precaución: for Research Use Only

Formula: C₆H₅NO₂
MW: 123.11 g/mol
Melting Pt: 139…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00006293
CAS Number: 98-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR