[Hydroxy(tosyloxy)iodo]benzene 96%

Supplier: Ambeed
Warning

Synonyms: Koser Reagent, Phenyliodosohydroxy Tosylate

A256827-100G A256827-10G A256827-25G A256827-5G A256827-500G
103852-154EA 6 USD
103852-154 76801-698 76801-700 77091-658 77304-690 77550-624
[Hydroxy(tosyloxy)iodo]benzene 96%
[Hydroxy(tosyloxy)iodo]benzene

Precaución: for Research Use Only

Formula: C₁₃H₁₃IO₄S
MW: 392.21 g/mol
Melting Pt: 131…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00011547
CAS Number: 27126-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR