Phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: Phenylmetaboric acid, Dihydroxy(phenyl)borane, Phenyl boric acid, Benzeneboronic acid

A111783-100G A111783-10G A111783-25G A111783-500G A111783-5G A111783-1000G
103808-076EA 161 USD
103808-076 103823-724 77108-014 77108-016 77108-018 77108-020 77284-180 77310-536
Phenylboronic acid 98%
Phenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BO₂
MW: 121.93 g/mol
Melting Pt: 218…222 °C
Flash Pt: 114.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00002103
CAS Number: 98-80-6
Merck Index: 13,01068

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR