4-Methylbenzophenone 95%

Supplier: Ambeed
Warning

Synonyms: p-Methylbenzophenone, Phenyl(p-tolyl)methanone

A105692-100G A105692-10G A105692-25G A105692-500G A105692-1000G
103807-938EA 57 USD
103807-938 77283-378 77283-380 77283-382 77283-384 77309-528
4-Methylbenzophenone 95%
4-Methylbenzophenone

Precaución: for Research Use Only

Formula: C₁₄H₁₂O
MW: 196.25 g/mol
Boiling Pt: 328.1 °C (760 mm Hg)
Melting Pt: 56.5…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00008553
CAS Number: 134-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR