2-(Trifluoromethyl)benzophenone 98%

Supplier: Ambeed
Warning

A189592-1G A189592-5G A189592-10G
103816-206EA 15 USD
103816-206 103846-800 77084-572 77084-574 77871-953
2-(Trifluoromethyl)benzophenone 98%
2-(Trifluoromethyl)benzophenone

Precaución: for Research Use Only

Formula: C₁₄H₉F₃O
Boiling Pt: 369 °C (760 mm Hg)
Melting Pt: 60…62 °C (760 mm Hg)
Flash Pt: 177 °C
Storage Temperature: Ambient
MDL Number: MFCD00000377
CAS Number: 727-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR