Phenyl acrylate 97% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

A456284-10G A456284-1G A456284-25G A456284-5G
103838-932EA 17 USD
103838-932 76725-644 76725-646 77307-400 77307-402
Phenyl acrylate 97% stabilized
Phenyl acrylate

Precaución: for Research Use Only

Formula: C₉H₈O₂
Boiling Pt: 232.2 °C (760 mm Hg)
Flash Pt: 88.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00048145
CAS Number: 937-41-7
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR