Phenethyl caffeate 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dihydroxycinnamic Acid Phenethyl Ester, Caffeic Acid Phenethyl Ester, Phenethyl 3-(3,4-dihydroxyphenyl)acrylate, CAPE, Phenethyl 3,4-Dihydroxycinnamate

A158279-100MG A158279-1G A158279-10G A158279-100G
103843-322EA 24 USD
103843-322 76689-686 76689-688 77625-470 77877-046
Phenethyl caffeate 98%
Phenethyl caffeate

Precaución: for Research Use Only

Formula: C₁₇H₁₆O₄
Storage Temperature: Freezer
MDL Number: MFCD00866470
CAS Number: 104594-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR