Phenazine-1-carboxamide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Oxychlororaphine
A146744-5G
A146744-1G
103803-696EA
227
USD
103803-696
76984-484
77299-544
Phenazine-1-carboxamide 98%
Phenazine-1-carboxamide
Precaución: for Research Use Only
Formula:
C₁₃H₉N₃O MW: 223.23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01708506 CAS Number: 550-89-0 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...