Pentafluorophenyl Acetate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Acetic acid Pentafluorophenyl ester
A112675-250MG
A112675-1G
103798-070EA
29
USD
103798-070
103823-908
76718-166
76718-168
77297-196
Pentafluorophenyl Acetate 98%
Pentafluorophenyl Acetate
Precaución: for Research Use Only
Formula:
C₈H₃F₅O₂ MW: 226.10 g/mol Boiling Pt: 187.4 °C (760 mm Hg) Melting Pt: 27 °C Flash Pt: 70.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00015552 CAS Number: 19220-93-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...